missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Porcelain Evaporating Dishes

Glazed inside and out except for rim.

Brand:  Fisherbrand™ FB968B

 View more versions of this product

Product Code. 13499719

  • 69.70 € / Pack of 6
Estimated Shipment: 30-12-2024
to see stock.
Explore more special offers
Add to basket

Description

Description

Porcelain dish with lip

  • Round bottom
  • Glazed inside and out except for rim.
  • Highly resistant to chemicals
  • Withstand temperatures up to 1150°C (2102°F)
  • Without cover

Specifications

Specifications

Round
Porcelain
Evaporating Dish
Glazed
1150°C
30 mm
2.95 in.
70 mL
Round
Evaporating Processes and Ashing
2102°F
2 mm
1.18 in.
75 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers