Learn More
Fisherbrand™ Powder Funnels
Parallel stems minimize bridging of powder.
Brand:  Fisherbrand™ 300700-0150
 View more versions of this product
Product Code. 11907914
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
Parallel stems minimize bridging of powder
  • Durable polypropylene
  • External ribbing prevents air locks
  • Autoclavable
TRUSTED_SUSTAINABILITY
Specifications
Specifications
15cm
27mm
13.7 cm
30mm
750 mL
Polypropylene
Autoclavable
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Fisherbrand™ Powder Funnels > Top dia. x H: 150 x 137mm; Capacity: 750mL