Learn More
Fisherbrand™ Powder Funnels
GREENER_CHOICE
Parallel stems minimize bridging of powder.
Brand:  Fisherbrand™ 300700-0080
117.55 EUR valid until 2025-04-30
Use promo code "24155" to get your promotional price.
 View more versions of this product
Product Code. 11907924
  • 155.00 € / Pack of 36
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
Parallel stems minimize bridging of powder
  • Durable polypropylene
  • External ribbing prevents air locks
  • Autoclavable
TRUSTED_SUSTAINABILITY
Specifications
Specifications
7.5cm
16mm
8.4 cm
29mm
100 mL
Polypropylene
Autoclavable