missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Clean-Wipes™

Cellulose and polyester blend cloth is soft and lint-free; will not scratch surfaces or fall apart like paper wipes.

Brand:  Fisherbrand™ 90023-31

28.37 EUR valid until 2025-12-31
Use promo code "24298" to get your promotional price.


 View more versions of this product

Product Code. 12972780

  • 31.40 € / Pack of 100

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers

Description

Description

Fisherbrand™ Clean-Wipes™ are ideal for cleaning or wiping optical parts, glassware, electrodes, cuvettes and microscopes.

 

  • Cellulose and polyester blend cloth is soft and lint-free; will not scratch surfaces or fall apart like paper wipes
  • Reclosable container dispenses single wipes, eliminates contamination from dust and dirt
  • Premoistened with 100% pure deionized water 

 

Order Info

Packed 100 wipes per polyethylene canister

Specifications

Specifications

Pre-moistened DI Wipes
22.8 cm
15.2 cm
9 x 6 in. (22.8 x 15.2 cm)
For cleaning or wiping optical parts, Glassware, electrodes, cuvettes and microscopes
Non-sterile
9 in.
6 in.
Polyester Cellulose
For cleaning or wiping optical parts, glassware, electrodes, cuvettes and microscopes.
100% pure deionized water
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Fisherbrand™ Clean-Wipes™ > Premoistened with deionized water

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.