missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Reusable Amber Glass Media Bottles
Click to view available options
Capacity (Metric):
10,000 mL
100 mL
1000 mL
2000 mL
250 mL
500 mL
5000 mL
Description
Fisherbrand™ Reusable Amber Glass Media Bottles with cap feature a convenient wide opening, enhanced graduations and a polypropylene pour ring. Feature clear (natural), drip-free, replaceable polypropylene pour ring.
- Permanent white enamel marking spot
- GL 45 screw thread
- Linerless
- Autoclavable (to 140°C)
- Amber borosilicate glass
- Polypropylene cap

Specifications
Specifications
Capacity (Metric) | 2000 mL |
Color | Amber |
For Use With (Application) | Media and reagent storage, Holding and dispensing fluids, Chemical storage |
Includes | Screw Cap, Pouring Ring |
Material | Borosilicate Glass (3.3) |
No. per Case | 1/Cs. |
Screw Color | Blue |
Screw Material | Polypropylene |
Sterility | Non-sterile |
Closure Color | Blue |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Fisherbrand™ Reusable Amber Glass Media Bottles >
Spot an opportunity for improvement?Share a Content Correction