missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Safety-Vented™ and Safety Labeled Right-to-Know Wash Bottles

Product Code. 12379195
Click to view available options
Capacity (Metric):
500 mL
Closure Color:
Assorted
Blue
Natural
Red
Yellow
For Use With (Application):
Acetone
Assorted (Acetone, Distilled Water, Ethanol, Isopropanol, Methanol, Sodium Hypochlorite)
Distilled Water
Ethanol
Isopropanol
Sodium Hypochlorite
This item is not returnable. View return policy
This item is not returnable. View return policy

Eliminates drips and allows bottle to dispense upright or inverted. Fisherbrand™ Safety-Vented™ and Safety Labeled Right-to-Know Wash Bottles' color-coded polypropylene closures aid in quick and easy identification.

  • Valve allows dispensing of wash bottle in an upright or inverted position
  • Wide mouth 53mm (2 in.) bottle for easy refilling
  • Translucent LDPE bottles with PP valves; Sodium Hypochlorite is white LDPE for light-protection between 230 and 450nm

Compliance

Provides compliance with OSHA requirements outlined in Hazard Communication Standard 29 CFR 1910.1200

Specifications

Capacity 16 oz., 500mL
Capacity (Metric) 500 mL
Capacity (English) 16 oz.
Color Yellow, Natural
For Use With (Application) Isopropanol
Material LDPE
Closure Color Yellow
Certifications/Compliance OSHA 29 CFR 1910.1200
Closure Included Yes
Autoclavable Not Autoclavable
Graduated No
Mouth Wide
Shape Cylindrical
Product Type Wash Bottle
Closure Material Polypropylene
Closure Size 53 mm
Closure Type Polypropylene Closure
Through the Closure Dispensing Yes
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'