Learn More
Fisherbrand™ Scientific Technical Pens
Fast-drying ink in four colors
Brand:  Fisherbrand™ 3046
 View more versions of this product
Product Code. 11724386
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
Fisherbrand™ Scientific Technical Pens produce extra-fine lines with accuracy—and without smearing. Special long-legged point is perfect for writing on notebooks, forms, graphs, charts, tables and recording paper. Fast-drying ink in four colors.
  • Ink is smooth flowing, fadeproof, and quick drying
  • Assortment includes red, blue, and special ultrablack for excellent reproduction on office copiers
  • Large ink reservoir lets you write for over 0.50 mile (800 m)
  • Cap design and color indicate point size and color of pen
  • Built-in super-tight cap seal keeps ink from drying out when pen is not in use
  • Choose from two line sizes: extra fine, 0.016 in. (0.4 mm), or super fine, 0.008 in. (0.2 mm)
  • Make precise lines with T-squares, rulers and flexible curves
Specifications
Specifications
Technical Pen
Single-use
6 Pk.
Blue
Used for record results in lab notebooks, fill in charts, draw graphs, mark recording paper, and produce diagrams
0.4 mm