missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LevGo™ Inoculation Picks
Isolate individual colonies with picks manufactured from White Birch wood. LevGo™ Inoculation Picks are naturally biodegradable and disposable, but are also autoclavable and suitable for food contact.
Brand: LevGo™ HS120011
Description
- Available in three lengths and two needle sizes to suit a wide range of application needs.
- Naturally biodegradable, made from 100% U.S.A grown white birch wood
- Manufactured in an FDA-registered factory and comply with FDA standards for food contact
- Available in semi or needle point styles for delicate operations
- Autoclavable
- Disposable
Specifications
Wooden Pick | |
Standard | |
1500 Pk. | |
63.5 mm |
White Birch Wood | |
Semi point | |
2.5 in. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction