Learn More
Fisherbrand™ Soil Analysis ASTM Hydrometers
These hydrometers are used to determine the specific gravity of soil in the range of 0.995 to 1.038 or -5 to 60g/L (using a weight by volume scale).
Brand:  Fisherbrand™ 13202132
 View more versions of this product
Product Code. 17112435
  • 81.00 € / Each
Estimated Shipment: 13-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
These hydrometers are used to determine the specific gravity of soil in the range of 0.995 to 1.038 or -5 to 60g/L (using a weight by volume scale).
  • 280mm (11 in.) long
  • Lead-free ballast and glass
  • Individually serialized
  • Conform to the accuracy requirements of ASTM hydrometers
  • Temperature standardization: 68°F
Storage:
Soil analysis hydrometers should be stored in a clean, dry, safe place, and should not be subjected to extreme temperatures.
Order Info
Should you require an individual Hydrometer Calibration Report, factory calibrated hydrometers or custom calibration services are available.
Certifications
Packaged with a multi-language (English, French, German, Italian, Portuguese, and Spanish) Statement of Accuracy indicating accuracy traceable to NIST. Hand Inspected by our U.S. quality team.
Specifications
Specifications
151H
0.001
280 mm
To determine the specific gravity of soil in the range of 0.995 to 1.038 or -5 to 60g/L (using a weight by volume scale)
±0.001
Traceable to NIST
Soil analysis hydrometers should be stored in a clean, dry, safe place, and should not be subjected to extreme temperatures.
0.995 to 1.038
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Fisherbrand™ Soil Analysis ASTM Hydrometers > ASTM 151H; Range: 0.995/1.038