missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Spilltray and Drying Rack

Polyethylene, chemical-resistant tray safely contains chemicals and liquids while pipetting, measuring or mixing

Brand:  Fisherbrand™ 90023-49

Product Code. 11704266

  • 64.25 € / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Description

Description

Polyethylene, chemical-resistant tray safely contains chemicals and liquids while pipetting, measuring or mixing

  • Holds up to 1.25L (42.5 oz.) of spilled liquid
  • Low profile, plastic grid design cushions and protects glassware and delicate items from breakage and scratches
  • Grid allows faster drying time by permitting air circulation
  • Easy to clean–dishwasher safe

Order Info

Traysize, 16.5L x 13W x 0.62 in. H (42 x 33 x 1.6cm)

Specifications

Specifications

Drying Rack
1.25 L Liquid
Chemicals and Liquids
Not Autoclavable
Plastic
16.5 x 13 x 0.625 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.