missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Stainless Steel Micro Spoons
Designed using high quality components these micro spoons feature a natural quality and bright finish.
107.00 € - 114.89 €
Description
- Designed using high quality components these micro spoons feature a natural quality and bright finish
- Laboratory Micro Spoon, stainless steel 304 spec 18/8 non-magnetic, bright finish
- Length = 100mm long
- Spoon measurement 8mm wide x 6mm long x 1.5mm deep
- Centre barrel diameter 1.60mm - 'micro'
- Spoon capacity [liquid] = 0.2ml
- Supplied in standard packs of 10
Specifications
Spoon | |
Stainless Steel |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Fisherbrand™ Stainless Steel Micro Spoons