Learn More
Fisherbrand™ Sterile Microcentrifuge Tubes with Screw Caps
GREENER_CHOICE
Note: Image is representative only. It shows colored caps, but all of the tubes in this offering come with Natural caps; there are no color cap options.
Brand:  Fisherbrand™ 515-S-FIS
105.11 EUR valid until 2025-12-31
Use promo code "24328" to get your promotional price.
 View more versions of this product
Product Code. 12350353
  • 168.00 € / Pack of 500
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
Choose skirted or conical-bottom style
  • With attached caps
  • Natural-colored screw caps with O-rings
  • RNase-free and DNase-free
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Polypropylene
1.5 mL
Polypropylene
Sterile
Conical
Autoclavable
Natural
O-ring
Screw Cap
30,000 x g
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Fisherbrand™ Sterile Microcentrifuge Tubes with Screw Caps > Capacity: 1.5mL; Conical bottom; Max. RCF: 30,000xG