missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Sterile Sampling Bags with Flat-Wire Closures

An economical and efficient way to collect, contain and carry samples

95.00 € - 131.00 €

Specifications

Material Polyethylene
View More Specs

Products 5
Product Code Brand Color Capacity (English) Thickness (English) Dimensions (L x W) Price Quantity & Availability  
Product Code Brand Color Capacity (English) Thickness (English) Dimensions (L x W) Price Quantity & Availability  
17903182
View Documents
Fisherbrand™
14955186
Clear 18 oz. 3 mil 9 x 4.5 in. (22.9 x 11.4cm)
95.00 €
Pack of 500
Please to purchase this item. Need a web account? Register with us today!
13158167
View Documents
Fisherbrand™
14955187
Clear 24 oz. 3.50 mil 9 x 5.5 in. (22.9 x 14cm)
131.00 €
Pack of 500
Estimated Shipment: 20-06-2025
to see stock.
Add to basket
 
11904405
View Documents
Fisherbrand™
14955190
Clear with White Block 24 oz. 3.50 mil 9 x 5.5 in. (22.9 x 14cm)
131.00 €
Pack of 500
Estimated Shipment: 20-06-2025
to see stock.
Add to basket
 
11914405
View Documents
Fisherbrand™
14955195
Clear with White Block 60 oz. 4 mil 10 x 10 in.
118.00 €
Pack of 250
Estimated Shipment: 20-06-2025
to see stock.
Add to basket
 
11738046
View Documents
Fisherbrand™
14955196
Clear 100 oz. 4 mil 14 x 10 in. (35.6 x 25.4cm)
122.00 €
Pack of 250
In Stock
to see stock.
Edge
Add to basket
 
Description

Description

Collect, contain and carry samples in an economical and efficient way

  • Sterile to avoid sample contamination
Specifications

Specifications

Polyethylene
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.