missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Traceable™ RTD Platinum Thermometer Replacement Probe

Traceable™ RTD Platinum Thermometer Replacement Probe for traceable RTD platinum.

Brand:  Fisherbrand™ 90028-84

Product Code. 12744078

  • 290.00 € / Each
Estimated Shipment: 04-03-2025
to see stock.
Explore more special offers
Add to basket

Description

Description

Traceable™ RTD Platinum Thermometer Replacement Probe for traceable RTD platinum.

  • 100 ohm
  • 8-wire probe
  • Probe length: 15.24 cm (6 in.)
Specifications

Specifications

152 mm
Thermometer
6 in.
For RTD platinum thermometer
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For improved performance, probes should be calibrated as a system with the meter.