missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Hostalen Tub for Ultrasonic Cleaners

The Fisherbrand™ Hostalen Tub is designed for the T 460, T 460/H, T 470/H and T 490 DH Ultrasonic Cleaners.

Brand:  Fisherbrand™ 2070400000

Product Code. 10065301

  • 22.55 € / Each
Estimated Shipment: 20-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Polypropylene material
  • Dimensions: 200 x 95 x 57mm
  • Two handles for ease-of-use
  • Solid bottom to keep parts secure
  • Immerse parts easily and safely

Specifications

Specifications

Hostalen Immersion Tub
Ultrasonic Cleaner, T 460, T 460/H, T 470/H and T 490 DH
Polypropylene
200 x 95 x 57 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.