missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Immersion Baskets for Ultrasonic Cleaners
This immersion basket is designed specifically for the Fisherbrand™ 112xx Series Dual Frequency Ultrasonic Cleaners. The fine mesh makes this immersion basket ideal for small items. The extra-long handle makes handling safe and easy.
16.40 € - 66.65 €
Specifications
Type | Immersion Basket |
---|---|
For Use With (Equipment) | FB11211 Ultrasonic Cleaning Unit |
Product Code | Brand | Material | Dimensions (L x W) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Material | Dimensions (L x W) | Price | Quantity & Availability | ||||
10196162
|
Fisherbrand™
1007545 |
Stainless Steel | 78 x 60 mm |
66.65 €
Each |
Estimated Shipment: 08-04-2025
Log in to see stock. |
||||
10611432
|
Fisherbrand™
2070380000 |
Polypropylene | 74 x 50 mm |
16.40 €
Each |
Estimated Shipment: 24-04-2025
Log in to see stock. |
||||
10089320
|
Fisherbrand™
2070540000 |
Stainless Steel | 59 x 60 mm |
52.30 €
Each |
Estimated Shipment: 24-04-2025
Log in to see stock. |
||||
Description
- Fine mesh for small part security
- Available in polypropylene or stainless steel
- Extra-long handle for ease-of-use
Specifications
Immersion Basket | |
FB11211 Ultrasonic Cleaning Unit |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title