Learn More
Fisherbrand™ Covers for Fisherbrand™ Ultrasonic Cleaners
A practical cover to complete your S-Line Ultrasonic Cleaning Unit. Fisherbrand™ Covers for S-Line Ultrasonic Cleaning Units are manufactured from durable blue plastic.
Brand:  Fisherbrand™ 1003278
 View more versions of this product
Product Code. 10060892
  • 14.90 € / Each
Estimated Shipment: 23-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Newly developed
  • Reduces noise and accelerates the heating-up process
  • When turned over, cover can be used as a drip-off basin
  • Collected cleaning liquid can be emptied back into the tank
Specifications
Specifications
Lid
Ultrasonic Cleaner, 0.8 L
Plastic