Learn More
Fisherbrand™ Covers for Fisherbrand™ Ultrasonic Cleaners
A practical cover to complete your S-Line Ultrasonic Cleaning Unit. Fisherbrand™ Covers for S-Line Ultrasonic Cleaning Units are manufactured from durable blue plastic.
Brand:  Fisherbrand™ 1003283
 View more versions of this product
Product Code. 11846244
  • 37.95 € / Each
Estimated Shipment: 27-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Easy-to-open and close cover
  • Muffles noise level
  • Designed to be used as drip basin when flipped over
  • Chemical-resistant plastic
  • Bring your ultrasonic cleaner up to temperature more quickly
  • Contain and prevent spills and leakage
Specifications
Specifications
Lid
Ultrasonic Cleaner, 6.9 and 9.4 L
Medical Laboratories and in Industrial Production Processes
Plastic