Learn More
Thermo Scientific™ FitBag™ UVM1 Broth
Shorten your workflow with easy to prepare 2.7L, 4.5L or 9L media bags: ready in 5 to 10 minutes, while segmented structure allows pre-filling.
1200.00 € - 1275.00 €
Specifications
Content And Storage | 10°C to 30°C |
---|---|
Description | FitBag UVM1 Broth |
Format | Convenience Format |
Product Type | Dehydrated Culture Media |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15705473
|
Thermo Scientific™
DF0863A |
30 x 2.7 L |
1220.00 €
Pack of 30 |
Estimated Shipment: 07-11-2024
Log in to see stock. |
|||||
15725473
|
Thermo Scientific™
DF0863C |
10 x 9.0 L |
1200.00 €
Pack of 10 |
Estimated Shipment: 07-11-2024
Log in to see stock. |
|||||
15715473
|
Thermo Scientific™
DF0863B |
20 x 4.5 L |
1275.00 €
Pack of 20 |
Estimated Shipment: 07-11-2024
Log in to see stock. |
|||||
Description
Save time in preparing a selective enrichment medium that conforms to the USDA-FSIS method for the isolation of Listeria monocytogenes, using Thermo Scientific™ FitBag™ UVM1 Broth. The irradiated medium is supplied in lightweight, transparent, plastic bags—just add water and the medium is ready for use. The bag's segmented structure allows pre-filling so that you can reconstitute in two minutes when needed. Available in 2.7L, 4.5L or 9L sizes to suit your workflow, medium can be dispensed through the outlet tube as required.
Shorten your workflow with FitBag UVM1 Broth. FitBag Enrichment Media is quick, easy-to-prepare, and highly adaptable to laboratory workflows and automated equipment, such as dilutors. No further supplementation is required.
Cost-effective: no need for weighing of media or sterilization, reducing the burden of media preparation, which frees up resources and equipment.
Flexible: segmented structure allows advanced preparation by filling one segment with deionized water from which FitBag Enrichment Media can be reconstituted within two minutes. Pre-sterilized water may also be added under aseptic conditions.
Easy-to-use: fill and dispense with minimum training.
Energy saving: gamma irradiated, eliminating the need for costly thermal processing.
Safe: eliminate handling of glassware, weighing of culture media, and the need to add supplementary reagents. Empty bags can be disposed of with normal laboratory waste.
Reduces waste: unprepared FitBag Enrichment Media can be kept for up to 24 months. Rehydrated media has a shelf life of 10 days.
Quality guaranteed: compliant with ISO 11133 and ISO 7218. Quality certificates are available for every batch, reducing the QC testing needed.
Space-saving: conveniently packed in flat boxes, FitBag Enrichment Media takes up relatively little space compared to prepared media and can be made up as required.
Specifications
10°C to 30°C | |
FitBag UVM1 Broth | |
Convenience Format | |
Dehydrated Culture Media |
Safety and Handling
missing translation for 'shelfLife' : 2 years (rehydrated 10 Days)