missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SCAT™ Fittings for HPLC Systems
Connect your HPLC capillaries safely and tightly to your SCAT™ system. SCAT™ PFA fittings are chemically resistant and fit Safety Caps and Safety Waste Caps. Available for different capillary sizes.
Brand: SCAT™ 107059
20.69 EUR valid until 2025-04-30
Use promo code "24211" to get your promotional price.
This item is not returnable.
View return policy
Description
SCAT fittings are available in the following versions:
- PFA fitting with integrated ferrule in various colors and diameters
- Tube connectors in various diameters
Specifications
Fitting | |
2.3 mm | |
Solvent Handling | |
Fitting with Integrated Ferrule (2.3 mm OD) | |
PFA |
Violet | |
2.3 mm | |
5 Fittings | |
Safety Caps, Safety Waste Caps, HPLC and UHPLC Systems |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
SCAT™ Fittings for HPLC Systems > Fittings; Color: Violet; For Use With: Safety Caps, Safety Waste Caps, HPLC and UHPLC Systems; Includes: Fitting with Integrated Ferrule (2.3 mm OD); Type: Fitting
Spot an opportunity for improvement?Share a Content Correction