Learn More
SCAT™ Fittings for HPLC Systems
Connect your HPLC capillaries safely and tightly to your SCAT™ system. SCAT™ PFA fittings are chemically resistant and fit Safety Caps and Safety Waste Caps. Available for different capillary sizes.
Brand:  SCAT™ 107906
31.10 EUR valid until 2024-12-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 14747170
  • 35.60 € / Pack of 10
Explore more special offers
Description
Description
SCAT fittings are available in the following versions:
  • PFA fitting with integrated ferrule in various colors and diameters
  • Tube connectors in various diameters
Specifications
Specifications
Ferrule
0.09 in.
Ferrule, 10 Pk.
PTFE
White
2.3 mm
Capillary