Learn More
SCAT™ Fittings for HPLC Systems
Connect your HPLC capillaries safely and tightly to your SCAT™ system. SCAT™ PFA fittings are chemically resistant and fit Safety Caps and Safety Waste Caps. Available for different capillary sizes.
Brand: SCAT™ 107167
This item is not returnable.
View return policy
Description
SCAT fittings are available in the following versions:
- PFA fitting with integrated ferrule in various colors and diameters
- Tube connectors in various diameters
Specifications
Plug | |
Solvent Handling | |
UNF 1/4 in. 28G | |
Polypropylene |
Colorless | |
5 x fitting 3.2 mm | |
SCAT Safety Caps |