missing translation for 'onlineSavingsMsg'
Learn More

Mettler Toledo™ FiveEasy™ F30 Conductivity Meters

Compact, benchtop meter provides high quality conductivity measurements with simple click of a button

Brand:  Mettler Toledo™ 30266943

983.09 EUR valid until 2024-12-31
Use promo code "21658" to get your promotional price.


 View more versions of this product

Product Code. 15513380

  • 1230.00 € / 1 unit
Estimated Shipment: 27-01-2025
to see stock.
Explore more special offers
Add to basket

Includes: LE703 sensor, installation QuickGuide, 1413μS/cm and 12.88mS/cm standard sachets
This item is not returnable. View return policy

Description

Description

  • Compact, space saving design with ABS housing
  • Remove sensor holder after use and store on the side of the meter
  • Large, well-structured 4.3 in. segmented LCD display for easy viewing
  • Intuitive button arrangement and simple menu for easy operation
  • One point calibration with three predefined standards
  • Mini-DIN sensor input
  • Features Automatic and manual endpoint recording, Acoustic endpoint signal, Visual endpoint signal
  • Ideal for wide range of laboratory applications
Specifications:
  • Conductivity Range: 0.01μS/cm to 200mS/cm
  • Conductivity Resolution: Automatic range
  • Conductivity Accuracy: ±0.5% of the measured value
  • TDS Range: 0.01mg/L to 200g/L
  • TDS Resolution: Automatic range
  • TDS Accuracy: ±0.5% of the measured value
  • Temperature Range: 0° to 100°C (32° to 212°F)
  • Temperature Resolution: 0.1°C
  • Temperature Accuracy: 0.5°C
  • Temperature Compensation: Linear: 0.00%/°C to 10.00%/°C, reference temperature: 20° and 25°C
  • Memory Size: current calibration
  • Power Supply: 100 to 240V, 50/60Hz, 12 VDC
  • Dimensions: 22.7 x 14.7 x 7cm (9 x 5.7 x 2.7 in.)
  • Weight: 0.63kg (1.4 lb.)
Applications:
  • F30 Meter only: Suited for applications that require a specific sensor not included in a kit
  • F30 Standard Kit: Suited for aqueous samples with mid to high conductivity
TRUSTED_SUSTAINABILITY
Specifications

Specifications

FiveEasy F30-Std
±0.5% of the measured value
One point calibration with three predefined standards
Automatic range
32°F to 212°F
0.5°C
Mini-DIN
0.63 kg
F30 Standard Kit
50/60Hz
F30 pH meter, LE703 pH sensor, electrode holder, 1 each buffers 1413mS/cm and 12.88mS/cm standard sachets, test certificate
Suited for aqueous samples with mid to high conductivity
0.01 μS/cm to 200 mS/cm
Linear: 0.00%/°C to 10.00%/°C, reference temperature: 20° and 25°C
0.1°C
Current calibration
22.7 x 14.7 x 7 cm (9 x 5.7 x 2.7 in.)
1.4 lb.
100/240V
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.