missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eppendorf™ Fixed Angle QuickLock™ Rotor with Aerosol-tight Lid
For use with Centrifuge 5430 and Centrifuge 5430 R
Brand: Eppendorf™ 5427754008
867.69 EUR valid until 2025-04-30
Use promo code "24119" to get your promotional price.
This item is not returnable.
View return policy
Description
- Aerosol-tight Eppendorf QuickLock rotors close with only 1/4 of a turn
- Saves time during every day use
- Takes repetitive stress off of your wrist

Specifications
Model: FA-45-48-11 | |
48 x 1.5 mL | |
12,700 rpm | |
Fixed | |
30 |
Fixed Angle Rotor | |
18,210 x g | |
5430/5430R Centrifuges | |
Aerosol rotor lid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Eppendorf™ Fixed Angle QuickLock™ Rotor with Aerosol-tight Lid > Model: FA-45-48-11
Spot an opportunity for improvement?Share a Content Correction