Learn More
Thermo Scientific™ Removable-Needle Autosampler Syringes for GC Instruments
Thermo Scientific™ Removable-Needle Autosampler Syringes for GC Instruments are accurate and reproducible over a range of volumes.
Brand:  Thermo Scientific™ 365C0971
 View more versions of this product
Product Code. 10299254
  • 88.35 € / Each
Explore more special offers
Description
Description
Thermo Scientific™ Removable-Needle Autosampler Syringes for GC Instruments are accurate and reproducible over a range of volumes.
  • Borosilicate glass with metal flanges
  • Needles and plungers are stainless steel
  • Field-proven syringe design
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Stainless Steel (Needle), Stainless Steel (Plunger)
23 to 26s G
GC Autosampler Syringe, Fixed Needle
Ea.
42 mm
Agilent GC Instruments
23 to 26 gauge; 5uL; 42mm length
42 mm
Reusable
5 μL
Cone
Fixed
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.