Learn More
Thermo Scientific™ Fixed-Needle, Super-Elastic Plunger Syringe
Thermo Scientific™ syringes for GC instruments are designed to provide enhanced durability,clarity and accuracy in sample introduction.
Brand:  Thermo Scientific™ 365D5416
70.80 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 15204573
  • 71.15 € / Each
Explore more special offers
Specifications
23 to 26 G
10 μL
Cone
GC Autosampler Syringe, Fixed Needle, Super-Elastic Plunger
42 mm
Agilent GC Instruments