Learn More
Macherey-Nagel™ Flash Chromatography Kit II
Brand:  Macherey-Nagel™ 727451
Product Code. 10566531
Quantity Price
1 703.00 € / Each
3 527.25 € / Each
Estimated Shipment: 23-04-2025
to see stock.
Explore more special offers
Add to basket
Specifications
Flash Chromatography Kit II
Includes One 450 x 40 mm I.D. Glass Column, One 1-L eluent Reservoir, 100 g Silica 60 (40 to 63 μm), Sea Sand, Silanized Glass Fiber Wadding, 1 m PTFE Tubing
Low Pressure Flash Chromatography
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Macherey-Nagel™ Flash Chromatography Kit II > Type: Flash Chromatography Kit II