Learn More
Paragon Scientific™ Flash Point Certified Reference Material
Flash Point Certified Reference Materials are for the verification of laboratory instruments used in testing petroleum and derivative products for Flash point according to ASTM D93.
Brand:  Paragon Scientific™ CRMU-PMGO
Product Code. 15480464
  • 215.00 € / 250mL
Explore more special offers
Description
Description
  • Certified Reference Material for annual verification checks in accordance with ASTM D93.
  • Fully traceable to international standards.
  • Supplied with MSDS.
  • Tamper evident security packaging.
  • 12 month shelf life.
  • Manufactured in the United Kingdom
Specifications
Specifications
MCC Diesel UKAS ISO 17034 Certified (56.2 °C), 250 mL
Certified Reference Material for annual verification checks in accordance with ASTM D93
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • CRMU-PMGO
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Aspiration toxicity Category 1
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P330-Rinse mouth.
  • P331-Do NOT induce vomiting.
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Nominal values quoted