Learn More
Paragon Scientific™ Flash Point Certified Reference Material
These are certified reference materials characterized by a method specific multi-laboratory study in accordance with ASTM D92. These materials are suitable for verifying the performance of an instrument and/or procedure as specified in this standard.
Brand:  Paragon Scientific™ CRMU-COC-MID
Product Code. 15862517
  • 320.00 € / Pack of 3
Explore more special offers
Description
Description
  • Certified Reference Material for annual verification checks in accordance with ASTM D92.
  • Certified according to ISO 17025 / ISO 17034 under UKAS accreditation by means of a method specific inter-laboratory study.
  • Packaged in 3 x 80 mL bottles – ‘One shot’, one bottle per test ensures zero contamination/ high-end volatile loss.
  • Fully traceable to international standards.
  • Supplied with MSDS.
  • Tamper evident security packaging.
  • 12 month shelf life.
  • Manufactured in the United Kingdom
Specifications
Specifications
COC ASTM D92 ( 153.0 °C), 3 x 80 mL
Certified Reference Material for annual verification checks in accordance with ASTM D92
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Show More
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Paragon Scientific™ Flash Point Certified Reference Material > COC ASTM D92 ( 153.0 °C), 3 x 80 mL
Nominal values quoted