missing translation for 'onlineSavingsMsg'
Learn More

Lonza™ FlashGel™ System DNA Cassettes

Cassettes optimized for DNA migration. Lonza™ FlashGel™ System DNA Cassettes separate DNA quickly and DNA migration can be seen as it happens. Separates DNA in 2 to 7 minutes.

261.00 € - 330.91 €

Specifications

Product Type DNA Cassettes
For Use With (Application) Electrophoresis
Kit Contents Agarose Gel, Stain and Buffer
Material Agarose
Quantity 9 Pack
View More Specs

Products 4
Product Code Brand Formulation Wells Separation Range Price Quantity & Availability  
Product Code Brand Formulation Wells Separation Range Price Quantity & Availability  
11597855
View Documents
Lonza™
57031
2.2% Agarose 12 + 1-well 10 to 1000 bp
330.91 €
Pack of 9
Please to purchase this item. Need a web account? Register with us today!
11527855
View Documents
Lonza™
57023
1.2% Agarose 12 + 1-well 50 to 4000 bp
322.00 €
Pack of 9
Estimated Shipment: 24-01-2025
to see stock.
Add to basket
 
11587855
View Documents
Lonza™
57029
1.2% Agarose 16 + 1-well 50 to 4000 bp This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11507865
View Documents
Lonza™
57032
2.2% Agarose 16 + 1-well 10 to 1000 bp
330.91 €
Each
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Get results in 5 minutes or less
  • Contain pre-stained agarose gels and buffer, minimizing user exposure to potentially mutagenic stains
  • For research or manufacturing use only
  • Not for use in in vitro diagnostic procedures, drug use, or for administration to humans or animals
Specifications

Specifications

DNA Cassettes
Agarose Gel, Stain and Buffer
9 Pack
4.5 in.
11.5 cm
4.2 in.
8.4 cm
1.7 cm
Electrophoresis
Agarose
18° to 26°C
2.7 in.
7 cm
10.7 cm
0.6 in.
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : 5 Months

Product Identifier
  • FlashGel System

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P264-Wash skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.

Supplemental information
  • MIXTURE LIST-Contains : Tetrabutylammonium hydroxide

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.