missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Micronova™ Flashlight Holder - Clamp
For use with Inspection Mirror to run the unit behind equipment and fixtures to ensure all surfaces have been properly cleaned
Brand: Micronova™ FLASHH-01
This item is not returnable.
View return policy
Description
- Use with the highly polished stainless steel mirror for inspecting hard to reach areas
- Features swivel connection allowing it to be attached to any SSU series handle
- The optional flashlight attachment is ideal for use in tanks and other low light applications
Specifications
Clamp style flashlight holder with 360° rotation | |
QDIST-66P (Inspection Mirror) | |
Flashlight Clamp |
3.6 x 2.0 x 1.2 in. | |
Plastic |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
1 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
032045-000 | Certificate of Compliance | 01/09/2022 | FLASHH-01 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Micronova™ Flashlight Holder - Clamp > Flashlight Holder (clamp)
Spot an opportunity for improvement?Share a Content Correction