Learn More
BUCHI FlashPure EcoFlex Flash Chromatography Cartridges: C18
FlashPure EcoFlex cartridges are designed to optimized backpressure, price and performance.
Brand: BUCHI 140000048
This item is not returnable.
View return policy
Description
- Most economical solution without sacrificing quality for standard separations
- Large selection of cartridge sizes from 4g-5,000g
- Non-RFID suitable on any system from BUCHI, ISCO, Biotage, Interchim or Yamazen
- Silica is ideal for basic applications and polar compounds using organic solvents
- Amino functionalized silica provides better peak shapes with polar and basic compounds and is ideal for carbohydrates
- C18 functionalized silica is ideal for peptides and non-polar compounds in an aqueous mobile phase
- Diol functionalized silica provides an alternate selectivity for a variety of polarity types and is ideal for lipids
- Neutral Alumina is ideal for aldehydes, ketones, and esters
Specifications
Polypropylene | |
5 Pc. | |
4 g | |
Non-Polar Compounds, Ideal for Peptides |
C18 | |
40 to 60 μm | |
C18 | |
Any Flash System |