Learn More
Labconco™ Flask Adapter
Suitable adapter for connecting 29/32 round-bottom flasks to valve ports on drying chambers and manifolds. Labconco™ Flask Adapter is constructed of stainless steel.
Brand: Labconco™ 7543800
Description
- Stainless steel construction
- Suitable for use with freeze dryers
Specifications
Stainless Steel | |
Freeze dryers | |
0.5 kg |
29/32 | |
3.5 in. wide, 0.75 in. diameter |
Product Suggestions
Customers who viewed this item also viewed
Product Content Correction
Product Title
Labconco™ Flask Adapter > Standard Joint No: 29/32
Spot an opportunity for improvement?Share a Content Correction