missing translation for 'onlineSavingsMsg'
Learn More

Scientific Industries Flask Clamp for Orbital Genie Shaker

Flask Clamp for Orbital Genie Shaker is a stainless steel flask clamp. Includes mounting screws.

Brand:  Scientific Industries SI-1615

 View more versions of this product

Product Code. 11391494

  • 102.00 € / Each
Estimated Shipment: 28-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Made of durable stainless steel
  • Two size options for either 250 mL flasks or 2 L flasks
  • Orbital Genie can accommodate twenty-five 250 mL flask clamps or five 2 L flask clamps
  • Includes mounting screws.
Specifications

Specifications

2 L Flasks
Stainless Steel
1 Ea.
Flask Clamp
Orbital Genie Shaker
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.