Learn More
Lenz Laborglasinstrumente™ Flat Flange Lid with Center Neck and Two Angled Side Necks
Brand:  Lenz Laborglasinstrumente™ 06103564
 View more versions of this product
Product Code. 11582354
  • 722.00 € / Each
Estimated Shipment: 05-02-2025
to see stock.
Explore more special offers
Add to basket
Specifications
Borosilicate Glass, Duran Glass
165 mm
Flange lid
Reaction vessels
Laboratory Flange 200