Learn More
Heidolph™ Flat Hood
For use as an add on to incubating shakers
Brand:  Heidolph™ 549-90040-00
Product Code. 10674131
  • 769.00 € / Each
Estimated Shipment: 28-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
With a height of 163 mm, the flat incubation hood is perfectly suited for multi-well plates, culture flasks and Erlenmeyer flasks with a volume up to 100 mL.
Specifications
Specifications
Multi-well plates, culture flasks and Erlenmeyer flasks with a volume up to 100 mL
Incubation Hood
163 mm
Incubating shakers