Learn More
Thermo Scientific™ Flat PCR Caps, strips of 12
Domed cap strips for PCR applications for use with Thermo Scientific 96-well PCR plates and strip tubes.
49.39 € - 95.25 €
Specifications
Color | Natural |
---|---|
Certifications/Compliance | Certified DNA/RNase Free |
Material | Polypropylene |
Product Code | Brand | Color | Product Type | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Product Type | Price | Quantity & Availability | ||||
12109840
|
Thermo Scientific™
AB0851 |
Natural | PCR Plate Cap Strip |
95.25 €
Pack of 200 |
Estimated Shipment: 09-10-2024
Log in to see stock. |
||||
12154681
|
Thermo Scientific™
AB0850 |
Natural | Flat Cap, 12-Cap Strip |
49.39 €
Pack of 80 |
Estimated Shipment: 25-10-2024
Log in to see stock. |
||||
Description
Ensure proper sealing on all PCR strip tubes and 96-well PCR plates with precision molded Thermo Scientific™ PCR Plate Cap Strips.
Flat PCR Cap Strips are compatible with 96-well PCR plates and standard and low profile strip tubes. The 12 strips have large end tabs for easy strip removal.Highlights:
- Suitable for use with all 96-well PCR plates as well as standard and low profile strip tubes
- Large end tabs for strip removal (12 strips only)
Certified free from human DNA, RNases, and DNases.
Especificaciones
Natural | |
Certified DNA/RNase Free | |
Polypropylene |