missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Flavopiridol hydrochloride, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Cyclin-dependent kinase (cdk) inhibitor (reported IC50 values are 6 - 25 nM and 84 - 200 nM for cdk9 and cdk2, respectively, and <0.85 μM for cdk1, cdk4, cdk5 and cdk7). Induces cell cycle arrest at G1 and G2 phase. Potently inhibits the growth of breast and lung cancer cell lines (IC50 = 25 - 160 nM) in vitro.
Specifications
Specifications
Quantity | 50 mg |
Formula Weight | 438.3 |
Percent Purity | >98% |
Chemical Name or Material | Flavopiridol hydrochloride |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'