missing translation for 'onlineSavingsMsg'
Learn More

Flecainide acetate, Tocris Bioscience™

Product Code. 18054932
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Cardiac Na+ channel blocker. Antiarrhythmic

Open Na+ channel blocker that inhibits fast Na+ current in cardiac muscle in a use- and concentration-dependent manner. Orally-active class Ic antiarrhythmic agent.

Chemical Identifiers

CAS 54143-56-5
Molecular Formula C19H24F6N2O5
Molecular Weight (g/mol) 474.4
InChI Key RKXNZRPQSOPPRN-UHFFFAOYSA-N
Synonym flecainide acetate, flecainide monoacetate, flecainide acetate salt, n-2-piperidinylmethyl-2,5-bis 2,2,2-trifluoroethoxy benzamide monoacetate, n-piperidin-2-ylmethyl-2,5-bis 2,2,2-trifluoroethoxy benzamide monoacetate, tambocor tn, 2,5-bis-2,2,2-trifluoroethoxy-n-2-piperidinylmethyl benzamide acetate, n-2-piperidylmethyl-2,5-bis 2,2,2-trifluoroethoxy benzamide monoacetate, benzamide, n-2-piperidinylmethyl-2,5-bis 2,2,2-trifluoroethoxy-, monoacetate
PubChem CID 41022
ChEBI CHEBI:5091
IUPAC Name acetic acid;N-(piperidin-2-ylmethyl)-2,5-bis(2,2,2-trifluoroethoxy)benzamide
SMILES CC(=O)O.C1CCNC(C1)CNC(=O)C2=C(C=CC(=C2)OCC(F)(F)F)OCC(F)(F)F

Specifications

Quantity 10 mg
Formula Weight 474.4
Percent Purity >99%
Chemical Name or Material Flecainide acetate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.