missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Flecainide acetate, Tocris Bioscience™
missing translation for 'orderingAttributeHoverText'
Quantity:
10 mg
50 mg
Description
Open Na+ channel blocker that inhibits fast Na+ current in cardiac muscle in a use- and concentration-dependent manner. Orally-active class Ic antiarrhythmic agent.
Specifications
Specifications
Quantity | 50 mg |
Formula Weight | 474.4 |
Percent Purity | >99% |
Chemical Name or Material | Flecainide acetate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'