missing translation for 'onlineSavingsMsg'
Learn More

Euro Protection™ Fleece Hat

Euro Protection™ Fleece Hat is constructed of thick, 300g/m2 polar fleece for warmth.

79.00 €

Specifications

Product Type Hat
Material Polar Fleece
Thickness 300 gsm
Includes Cord for tightening
View More Specs

Products 1
Product Code Brand Color Price Quantity & Availability  
Product Code Brand Color Price Quantity & Availability  
15804105
View Documents
Euro Protection™
5COVEO
Orange
79.00 €
Pack of 10
Estimated Shipment: 21-01-2025
to see stock.
Add to basket
 
Description

Description

  • Constructed of 100% polyester polar fleece,300g/m2
  • Includes black tightening cord for secure fit
Specifications

Specifications

Hat
Polar Fleece
300 gsm
Cord for tightening
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.