Learn More
Eppendorf™ Flex Bucket Adapters for IsoRack
Add flexibility to your liquid handling applications with these IsoRack adapters
164.66 € - 173.69 €
Specifications
Type Adapter
For Use With (Equipment) Flex bucket
No. of Places 96
No. per Pack 2 Pack
View More Specs
Products 2
Product Code Brand Capacity Price Quantity & Availability  
Product Code Brand Capacity Price Quantity & Availability  
10224032
View Documents
Eppendorf™
5825708008
0.5 mL
164.66 €
Pack of 2
10171362
View Documents
Eppendorf™
5825709004
1.5 to 2 mL
173.69 €
Pack of 2
Description
Description
  • IsoRack adapters to support 0.5 mL and 1.5/2.0 mL IsoRacks during centrifugation
  • Adapters can be used in Eppendorf plate rotors A-2-DWP and A-4-81- MTP/Flex
Specifications
Specifications
Adapter
Flex bucket
96
2 Pack