Learn More
Digi-Sense™ Flexible Insulated-Wire Thermocouple Probes
Optimize readings and temperature accuracy for a variety of applications with these probes. Cole-Parmer™ Digi-Sense™ Flexible Insulated-Wire Thermocouple Probes are ideal for high-temperature measurements and can be used with liquids, gases and semisolids.
37.85 € - 107.00 €
Specifications
Certifications/Compliance | ANSI |
---|---|
For Use With (Equipment) | Temperature Meters |
Type | Thermocouple Probe |
Product Code | Brand | Temperature (Metric) | Style | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Temperature (Metric) | Style | Price | Quantity & Availability | ||||
11860000
|
Digi-Sense™
08516-65 |
-150°C to +204°C | Type-K, Std Pen Probe; 4 in., Mini Conn |
76.00 €
Each |
Estimated Shipment: 29-04-2025
Log in to see stock. |
||||
11849652
|
Digi-Sense™
08439-62 |
-200°C to +311°C | Compact Thermocouple Probe, 4.5 in., Grnd; Type K |
49.80 €
Each |
Estimated Shipment: 24-04-2025
Log in to see stock. |
||||
11801375
|
Digi-Sense™
08514-66 |
-150°C to +649°C | Type-K Comp Surface Probe 4.5 in. Mini Conn Exp 5 ft. Fiberglass / SS Cable |
71.00 €
Each |
Estimated Shipment: 23-06-2025
Log in to see stock. |
||||
11763099
|
Digi-Sense™
08516-71 |
-150°C to +1100°C | Type-K Pipe-Ftg Probe SS Handle Mini Conn |
60.10 €
Each |
Estimated Shipment: 15-05-2025
Log in to see stock. |
||||
11850000
|
Digi-Sense™
08516-55 |
-200°C to +311°C | Type-K, Standard Probe, Mini Conn, 5 in., .093 Dia Tip, GRD, 5 ft. Coil Cord |
69.10 €
Each |
Estimated Shipment: 24-04-2025
Log in to see stock. |
||||
11781349
|
Digi-Sense™
08467-64 |
-150°C to +1093°C | Type-K High-Temp Ceramic Fiber-Insulated Probe Mini Conn Exp |
55.25 €
Each |
Estimated Shipment: 29-04-2025
Log in to see stock. |
||||
11871435
|
Digi-Sense™
08469-22 |
-150°C to +482°C | Type-K Hose Clamp Probe Mini; Conn GRD 10ft. SS Braid Cable |
37.90 €
Each |
Estimated Shipment: 23-06-2025
Log in to see stock. |
||||
11880000
|
Digi-Sense™
08516-96 |
-250°C to +1100°C | Type-K High-temperature Flexible thermocouple probe;12 in. |
49.85 €
Each |
Estimated Shipment: 24-04-2025
Log in to see stock. |
||||
11809662
|
Digi-Sense™
08439-82 |
-150°C to +148°C | Compact Penetration Thermocouple Probe, 2.5 in., Grnd; Type K |
49.85 €
Each |
Estimated Shipment: 06-06-2025
Log in to see stock. |
||||
11830010
|
Digi-Sense™
08117-45 |
-150°C to +427°C | Detachable Thermocouple Probe, 4 in., Penetration, Grnd; Type K |
37.85 €
Each |
Estimated Shipment: 06-06-2025
Log in to see stock. |
||||
11771339
|
Digi-Sense™
08516-74 |
-150°C to +482°C | Type-K Pipe Plug Probe SS 1 / 4 in. NPT(M) Mini Conn |
60.10 €
Each |
Estimated Shipment: 24-04-2025
Log in to see stock. |
||||
11735249
|
Digi-Sense™
08469-82 |
-150°C to +100°C | Compact Penetration Thermocouple Probe, 2.5 in. L, Grnd; Type K |
62.75 €
Each |
Estimated Shipment: 23-06-2025
Log in to see stock. |
||||
11752683
|
Digi-Sense™
08516-97 |
-250°C to +1100°C | Thermocouple Type: K |
107.00 €
Each |
Estimated Shipment: 24-04-2025
Log in to see stock. |
||||
Description
Optimize readings and temperature accuracy for a variety of applications with these probes. Cole-Parmer™ Digi-Sense™ Flexible Insulated-Wire Thermocouple Probes are ideal for high-temperature measurements and can be used with liquids, gases and semisolids.
All insulated-wire probes include straight insulated cable with no handle. Fiberglass and ceramic insulated probes have exposed junctions; use for high-temperature applications. Exposed junction probes are not recommended for use with aqueous solutions (type J exposed-junction probes will rust when exposed to moisture).- Each probe comes with color coded ANSI mini-connectors: type J-black, type K-yellow, type T-blue, and type E-purple.
- Ceramic Fiber-Insulated Wire Probes for high-temperature measurements.
- General-Purpose Probes are designed for use with liquids, gases and semisolids.
- ABS handles provide a light weight design for ease of use along with durability
- Probe handles feature a strain relief spring that protects the cable connection against damage due to repeated flexing and tugging
Specifications
ANSI | |
Temperature Meters | |
Thermocouple Probe |