Learn More
STEMCELL Technologies Single Flow Meter
Meter to control gas flow into hypoxia incubator chamber
Brand:  STEMCELL Technologies 27311
Encompass_Preferred
Product Code. 16231962
  • 785.70 € / Each
Estimated Shipment: 09-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Single Flow Meter allows for precise control of gas flow to generate a hypoxic environment for tissue culture using the Hypoxia Incubator Chamber (Catalog #27310)
  • Features of the Single Flow Meter include:
  • Same precision as the flow meter used for full size continuous flow carbon dioxide incubator chambers
  • Calibrated for 5 - 50 liters per minute (LPM)
Specifications
Specifications
Cell Culture
1 m