missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ FLoid™ Cell Imaging Station

Affordable, user-friendly imaging solution for quick detection and verification of fluorescently-labeled samples

Brand:  Invitrogen™ 4471136

13523.84 EUR valid until 2024-11-29
Use promo code "21683" to get your promotional price.


Product Code. 15520390

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers

Requires : Printer
This item is not returnable. View return policy

Description

Description

Uses an open-stage design providing maximum vessel flexibility for slides, coverslips, multi-well culture dishes, or even T75 flasks. Captures transmitted light and three-color fluorescent images of cells and samples at benchtop without special training or dedicated staff. Simple user interface, streamlined image acquisition process, and real-time, multicolor display allow users to produce high-quality images with a few mouse clicks.

  • Simplicity: Fully integrated system with intuitive, multilingual user interface
  • Speed: Get results in a snap, without warm-up, cool-down, or filter changes
  • Convenience: Capture and print images on your bench, rather than in the darkroom
  • Information: Provides information on Molecular Probes™ reagents and protocols via user interface
  • Robustness: No moving parts and long life LEDs for reliable day to day use
  • High Quality Imaging Anywhere in Lab
  • Versatile and easy to use
  • Fast and reliable
  • An everyday use imager for every user

System specifications

  • Objective: Fixed 20X plan fluorite; numerical aperture (NA)= 0.45; working distance (WD)=5.9 mm
  • Color Channels: 4 channels (relief phase, blue, green, and red fluorescence)
  • Illumination: LED (50,000 hour life), adjustable intensity
  • Excitation: Blue channel: 390/40nm; Green channel: 482/18nm; Red channel: 586/15nm
  • Emission: Blue channel: 446/33nm; Green channel: 532/59nm; Red channel: 646/68nm
  • Captured Images: 16-bit monochrome, TIFF, PNG, JPG, or BMP (129X 964 pixels)
  • LCD display: 15 in. color; display resolution 1366 x 768 pixels; image resolution 1296 x 964 pixels; adjustable tilt

Cell Analysis, Cellular Imaging, Immunofluorescence (IF), Immunofluorescence Staining & Detection

Order Info

W x H x D: (40 x 54 x 35cm)16 x 21 x 14 in. Weight: 12.7kg (28 lb.). Shipping Condition: Room Temperature.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Cell Imaging Station
54cm
35cm
40cm
21 in.
14 in.
16 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers