Learn More
Scotch-Brite™ Floor Pads, Green
For the efficient, intermediate cleaning of heavily soiled floors with or without a coating. For use with single disc machines or automatic floor cleaning machines with slow speed.
Brand:  Scotch-Brite™ 7100159882
 View more versions of this product
Product Code. 17128810
  • 511.00 € / Pack of 25
Estimated Shipment: 21-03-2025
to see stock.
Explore more special offers
Add to basket
Specifications
Floor Pad
460 mm
Single Disc Machines or Slow Speed Automatic Floor Cleaning Machines
Green
Scrubbing
Round