Learn More
Scotch-Brite™ Floor Pads, Red
Designed for everyday cleaning and light scrubbing to remove light soil, scuff marks, and black heel marks. These pads clean when damp and buff when dry. Open-web construction means this washable, reusable pad resists clogging and loading.
Brand:  Scotch-Brite™ 7100182731
 View more versions of this product
Product Code. 17128850
  • 116.00 € / Pack of 5
Estimated Shipment: 21-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • For everyday cleaning and light scrubbing
  • Removes light soil, scuff marks, and black heel marks
  • Cleans when damp and buffs when dry
  • Conformable fibers provide maximum pad-to-floor contact
  • Reversible, washable pad resists clogging and loading
  • Consistent, proven performance with long floor pad life
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Floor Pad
Red
480 mm
Everyday Cleaning, Light Scrubbing