Learn More
Scotch-Brite™ Floor Pads, White
Ideal for dry polishing and buffing unprotected floors with standard speed rotary floor machines. These pads are also suitable for light spray cleaning. They enhance floor shine without scratching or damaging protected floors.
Brand:  Scotch-Brite™ 7100159897
 View more versions of this product
Product Code. 17168820
  • 90.00 € / Pack of 5
Estimated Shipment: 21-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Used for light cleaning to help remove soil and scuff marks with minimal dulling of finish
  • Highly conformable fibers provide maximum pad-to-floor contact, delivering outstanding cleaning results
  • Reversible and washable polishing pad resists clogging and loading
  • Uniform distribution of mineral abrasives throughout whole pad and not just on the surface
  • Delivers consistent performance with long floor pad life
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Floor Pads
432 mm