missing translation for 'onlineSavingsMsg'
Learn More

Flow Cup Viscosity Standards, Paragon Scientific™

Product Code. p-7161069
Click to view available options
Viscosity:
1044 mPa.s (20°C)
135.9 mPa.s (20°C)
17.78 mPa.s (20 °C)
1882 mPa.s (20 °C)
281.4 mPa.s (20°C)
37.22 mPa.s (20°C)
557.7 mPa.s (20°C)
72.9 mPa.s (20°C)
8.945 mPa.s (20°C)
Unit Size:
500mL
This item is not returnable. View return policy

Product Code. 16050172

Brand: Paragon Scientific C6

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

These standards are especially designed for use in DIN, Ford, ISO, Shell and Zahn flow cups. Certified at 20°C and 25°C, each value is tested to ASTM D2162 for viscosity measurement and ASTM D1480 for Density measurement, certified to the International Standards BS EN ISO / IEC 17025 and ISO 17034.

  • Certified according to ISO 17025 / ISO 17034 under UKAS accreditation
  • Manufactured in accordance with ASTM D2162 the primary method for viscosity
  • Density g/mL in accordance with ASTM D1480
  • Density, dynamic viscosity and kinematic viscosity quoted at all temperatures
  • Ensures full compliance to ASTM & IP test method protocol
  • Full Traceability to National Standards
  • 2 year shelf life

Specifications

Viscosity 8.945 mPa.s (20°C)
Quantity 500 mL
compliance-icons
Product Identifier
  • 500ML C6 Flow Cup Viscosity Standard
Signal Word
  • Danger
Hazard Category
  • Aspiration toxicity Category 1
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
Precautionary Statement
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.

Nominal values quoted

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.