missing translation for 'onlineSavingsMsg'
Learn More

Kartell™ Flow Indicator Ball

Ideal method of monitoring fluid direction, flow, and color. Kartell™ Flow Indicator Ball can be used horizontally or vertically and the speed of the ball is proportional to the flow.

Brand:  Kartell™ 0053800

Product Code. 11318773

  • 22.05 € / Each
Estimated Shipment: 23-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • For liquid flow from 50mL/min. in horizontal position
  • For liquid flow from 140mL/min. in vertical position
  • Minimum air flow 3L/min. horizontal
  • Minimum air flow 8L/min. vertical
  • Speed of rotation of ball proportional to flow
  • Not to be used with corrosive fluids or over 50°C
  • Connector has O.D. of 6.5 - 10mm
Specifications

Specifications

Flow Ball Indicator
For liquid flow from 50mL/min. in horizontal position and 140mL/min, in vertical position
102 mm
50 mL/min.
57 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.